Learn More
Biomatik Corporation Human Excitatory Amino Acid Transporter 1 (EAAT1) ELISA Kit
Supplier:  Biomatik Corporation EKU03989
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.125ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: SLC1A3; EA6; GLAST1; Sodium-dependent glutamate/aspartate transporter 1; Solute Carrier 1 Member 3,Glial High Affinity Glutamate Transporter; GLutamate Aspartate Transporter. Restriction: For Research Use Only.
Catalog No. 50-228-5992