Learn More
Biomatik Corporation Mouse Family With Sequence Similarity 3, Member B (FAM3B) ELISA Kit
Supplier:  Biomatik Corporation EKU04019
Encompass_Preferred
Mouse Family With Sequence Similarity 3, Member B (FAM3B) ELISA Kit for detecting FAM3B in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9D309. Target Synonyms: ORF9; PANDER; PRED44; C21orf11; C21orf76; Cytokine-like protein 2-21; Pancreatic-derived factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5994