Learn More
Biomatik Corporation Human GA Binding Protein Transcription Factor Alpha (GABPa) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04307
Encompass_Preferred
The Human GA Binding Protein Transcription Factor Alpha (GABPa) ELISA Kit allows for quantitative determination of GA Binding Protein Transcription Factor Alpha in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.061 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q06546. Cross Activity: No significant cross-reactivity or interference between GA Binding Protein Transcription Factor Alpha and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2548