Learn More
Biomatik Corporation Rat Fibrinogen Degradation Product (FDP) ELISA Kit
Supplier:  Biomatik Corporation EKU04101
Encompass_Preferred
Rat Fibrinogen Degradation Product (FDP) ELISA Kit for detecting FDP in plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.8-500pg/ml. Sensitivity: 3.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Target Synonyms: FnDP; Fibrin Split Products; Fibrin Degradation Product. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6004