Learn More
Biomatik Corporation Human Galanin (GAL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04311
Encompass_Preferred
The Human Galanin (GAL) ELISA Kit allows for quantitative determination of Galanin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 12.35-1,000 pg/mL. Sensitivity: 4.92 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P22466. Cross Activity: No significant cross-reactivity or interference between Galanin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2551