Learn More
Biomatik Corporation Human Galectin 1 (GAL1) ELISA Kit, 96 tests
Catalog No. 501502552
Encompass_Preferred
The Human Galectin 1 (GAL1) ELISA Kit allows for quantitative determination of Galectin 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.123 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P09382. Cross Activity: No significant cross-reactivity or interference between Galectin 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2552 Supplier Biomatik Corporation Supplier No. EKU04314
The Human Galectin 1 (GAL1) ELISA Kit allows for quantitative determination of Galectin 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.123 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P09382. Cross Activity: No significant cross-reactivity or interference between Galectin 1 and analogues was observed. Restriction: For Research Use Only.