Learn More
Biomatik Corporation Human Furin (FUR) ELISA Kit
Supplier:  Biomatik Corporation EKU04288
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P09958. Target Synonyms: PACE; PCSK3; SPC1; Paired Basic Amino Acid Cleaving Enzyme; Dibasic-processing enzyme. Restriction: For Research Use Only.
Catalog No. 50-228-6024