Learn More
Biomatik Corporation Mouse Gastric Inhibitory Polypeptide (GIP) ELISA Kit
Supplier:  Biomatik Corporation EKU04341
Encompass_Preferred
Mouse Gastric Inhibitory Polypeptide (GIP) ELISA Kit for detecting GIP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 61.7-5,000pg/ml. Sensitivity: 26.1pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P48756. Gene ID: 14607. Target Synonyms: Incretin hormone; Glucose Dependent Insulinotropic Peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6027