Learn More
Biomatik Corporation Pig Ghrelin (GHRL) ELISA Kit
Supplier:  Biomatik Corporation EKU04364
Encompass_Preferred
Pig Ghrelin (GHRL) ELISA Kit for detecting GHRL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 123.5-10,000pg/ml. Sensitivity: 50.7pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q9GKY5. Gene ID: 396728. Target Synonyms: MTLRP; Growth Hormone-Releasing Peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6030