Learn More
Biomatik Corporation Rat Glial Cell Line Derived Neurotrophic Factor (GDNF) ELISA Kit
Supplier:  Biomatik Corporation EKU04377
Encompass_Preferred
Rat Glial Cell Line Derived Neurotrophic Factor (GDNF) ELISA Kit for detecting GDNF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78-5,000pg/ml. Sensitivity: 28pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q07731. Gene ID: 25453. Target Synonyms: ATF1; ATF2; HFB1-GDNF; Astrocyte-Derived Trophic Factor; Glial Derived Neurotrophic Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6032