Learn More
Biomatik Corporation Human Gastric Inhibitory Polypeptide (GIP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04340
Encompass_Preferred
The Human Gastric Inhibitory Polypeptide (GIP) ELISA Kit allows for quantitative determination of Gastric Inhibitory Polypeptide in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 61.7-5000 pg/mL. Sensitivity: 23.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P09681. Cross Activity: No significant cross-reactivity or interference between Gastric Inhibitory Polypeptide and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2563