Learn More
Biomatik Corporation Human Chitinase-3-like Protein 1 (CHI3L1) ELISA Kit
Supplier:  Biomatik Corporation EKU04527
Encompass_Preferred
Human Chitinase-3-like Protein 1 (CHI3L1) ELISA Kit for detecting CHI3L1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4,000pg/ml. Sensitivity: 23.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P36222. Gene ID: 1116. Target Synonyms: GP39; CHI3-L1; YKL40; Chondrex; Chitinase 3-Like 1; Chondrocyte Protein YKL40; 39 kDa synovial protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6046