Learn More
Biomatik Corporation Dog Gastrin (GT) ELISA Kit, 96 tests
Catalog No. 501502565
Encompass_Preferred
The Dog Gastrin (GT) ELISA Kit allows for quantitative determination of Gastrin in Serum, Plasma and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 12.35-1000 pg/mL. Sensitivity: 4.95 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P01353. Cross Activity: No significant cross-reactivity or interference between Gastrin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2565 Supplier Biomatik Corporation Supplier No. EKU04344
The Dog Gastrin (GT) ELISA Kit allows for quantitative determination of Gastrin in Serum, Plasma and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 12.35-1000 pg/mL. Sensitivity: 4.95 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P01353. Cross Activity: No significant cross-reactivity or interference between Gastrin and analogues was observed. Restriction: For Research Use Only.