Learn More
Biomatik Corporation Human Gastrin (GT) ELISA Kit, 96 tests
Catalog No. 501502566
Encompass_Preferred
The Human Gastrin (GT) ELISA Kit allows for quantitative determination of Gastrin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 12.35-1,000 pg/mL. Sensitivity: 4.39 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P01350. Cross Activity: No significant cross-reactivity or interference between Gastrin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2566 Supplier Biomatik Corporation Supplier No. EKU04346
The Human Gastrin (GT) ELISA Kit allows for quantitative determination of Gastrin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 12.35-1,000 pg/mL. Sensitivity: 4.39 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P01350. Cross Activity: No significant cross-reactivity or interference between Gastrin and analogues was observed. Restriction: For Research Use Only.