Learn More
Biomatik Corporation Mouse Gremlin 1 (GREM1) ELISA Kit
Supplier:  Biomatik Corporation EKU04573
Encompass_Preferred
Mouse Gremlin 1 (GREM1) ELISA Kit for detecting GREM1 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O70326. Gene ID: 23892. Target Synonyms: PIG2; CKTSF1B1; DAND2; IHG-2; DAND2; DRM; Increased in high glucose protein 2; Cell proliferation-inducing gene 2 protein; Cysteine knot superfamily 1, BMP antagonist 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6051