Learn More
Biomatik Corporation Rat Gastrin (GT) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04348
Encompass_Preferred
Rat Gastrin (GT) ELISA Kit for detecting GT in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.54pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P04563. Gene ID: 25320. Target Synonyms: GAST; GAS; Gastrin component I; Big gastrin; Gastrin component II; Gastrin component III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2568