Learn More
Biomatik Corporation Horse Growth Hormone (GH) ELISA Kit
Supplier:  Biomatik Corporation EKU04612
Encompass_Preferred
Horse Growth Hormone (GH) ELISA Kit for detecting GH in serum, plasma, other biological fluids. Species Reactivity: Horse (Equus caballus; Equine). Detection Range: 246.9-20,000pg/ml. Sensitivity: 91.3pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01245. Gene ID: 100034180. Target Synonyms: GH1; GH-N; GHN; hGH-N; Somatotropin; Hygetropin; Jintropin; Kigtropin; Pituitary Growth Hormone; Growth Hormone, Normal. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6057