Learn More
Biomatik Corporation Human GATA Binding Protein 3 (GATA3) ELISA Kit, 96 tests
Catalog No. 501502569
Encompass_Preferred
The Human GATA Binding Protein 3 (GATA3) ELISA Kit allows for quantitative determination of GATA Binding Protein 3 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2000 pg/mL. Sensitivity: 11.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P23771. Cross Activity: No significant cross-reactivity or interference between GATA Binding Protein 3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2569 Supplier Biomatik Corporation Supplier No. EKU04354
The Human GATA Binding Protein 3 (GATA3) ELISA Kit allows for quantitative determination of GATA Binding Protein 3 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2000 pg/mL. Sensitivity: 11.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P23771. Cross Activity: No significant cross-reactivity or interference between GATA Binding Protein 3 and analogues was observed. Restriction: For Research Use Only.