Learn More
Biomatik Corporation Human Gelsolin (GS) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04357
Encompass_Preferred
The Human Gelsolin (GS) ELISA Kit allows for quantitative determination of Gelsolin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 62.5-4000 pg/mL. Sensitivity: 28.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P06396. Cross Activity: No significant cross-reactivity or interference between Gelsolin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2570