Learn More
Biomatik Corporation Chicken Heat Shock Protein 70 (HSP70) ELISA Kit
Supplier:  Biomatik Corporation EKU04669
Encompass_Preferred
Chicken Heat Shock Protein 70 (HSP70) ELISA Kit for detecting HSP70 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08106. Gene ID: 423504. Target Synonyms: DnaK; 70 Kilodalton Heat Shock Proteins. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6062