Learn More
Biomatik Corporation Human Ghrelin (GHRL) ELISA Kit, 96 tests
Catalog No. 501502571
Encompass_Preferred
The Human Ghrelin (GHRL) ELISA Kit allows for quantitative determination of Ghrelin in Serum, Plasma, Tissue homogenates, Cell lysates, Saliva, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 123.5-10,000 pg/mL. Sensitivity: 49.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: Q9UBU3. Cross Activity: No significant cross-reactivity or interference between Ghrelin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2571 Supplier Biomatik Corporation Supplier No. EKU04361
The Human Ghrelin (GHRL) ELISA Kit allows for quantitative determination of Ghrelin in Serum, Plasma, Tissue homogenates, Cell lysates, Saliva, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 123.5-10,000 pg/mL. Sensitivity: 49.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: Q9UBU3. Cross Activity: No significant cross-reactivity or interference between Ghrelin and analogues was observed. Restriction: For Research Use Only.