Learn More
Biomatik Corporation Mouse Hexosaminidase B Beta (HEXb) ELISA Kit
Supplier:  Biomatik Corporation EKU04777
Encompass_Preferred
Mouse Hexosaminidase B Beta (HEXb) ELISA Kit for detecting HEXb in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P20060. Gene ID: 15212. Target Synonyms: Hex-B; ENC-1AS; HCC-7; Beta-N-acetylhexosaminidase subunit beta; Cervical cancer proto-oncogene 7 protein; N-acetyl-beta-glucosaminidase subunit beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6070