Learn More
Biomatik Corporation Rat High Density Lipoprotein (HDL) ELISA Kit
Supplier:  Biomatik Corporation EKU04782
Encompass_Preferred
Rat High Density Lipoprotein (HDL) ELISA Kit for detecting HDL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.47ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04639. Gene ID: 25081. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6073