Learn More
Biomatik Corporation Homovanillic Acid (HVA) ELISA Kit
Supplier:  Biomatik Corporation EKU04835
Encompass_Preferred
Homovanillic Acid (HVA) ELISA Kit for detecting HVA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 2.47-200ng/ml. Sensitivity: 0.95ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: Homovanilic Acid; HOC6H3 (OCH3) CH2COOH; 3-Methoxy-4-hydroxyphenyl Acetic Acid; 4-Hydroxy-3-Methoxy-Benzeneacetic Acid; 4-Hydroxy-3-Methoxyphenylacetic Acid. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6075