Learn More
Biomatik Corporation Human Glial Cell Line Derived Neurotrophic Factor (GDNF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04375
Encompass_Preferred
The Human Glial Cell Line Derived Neurotrophic Factor (GDNF) ELISA Kit allows for quantitative determination of Glial Cell Line Derived Neurotrophic Factor in Serum, Plasma, Tissue homogenates, Cell lysates, Cerebrospinal fluid, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.058 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P39905. Cross Activity: No significant cross-reactivity or interference between Glial Cell Line Derived Neurotrophic Factor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2576