Learn More
Biomatik Corporation Dog Immunoglobulin G (IgG) ELISA Kit
Supplier:  Biomatik Corporation EKU04890
Encompass_Preferred
Dog Immunoglobulin G (IgG) ELISA Kit for detecting IgG in serum, plasma, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 12.35-1,000ug/ml. Sensitivity: 5.21ug/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6086