Learn More
Biomatik Corporation Rat Immunoglobulin G1 (IgG1) ELISA Kit
Supplier:  Biomatik Corporation EKU04902
Encompass_Preferred
Rat Immunoglobulin G1 (IgG1) ELISA Kit for detecting IgG1 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 49.4-4,000ng/ml. Sensitivity: 18.7ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P20759. Target Synonyms: IGHG1; Ig Gamma-1 Chain C Region; Immunoglobulin Heavy Constant Gamma 1; G1m Marker; Immunoglobulin Gm1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6090