Learn More
Biomatik Corporation Human Glucagon (GC) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04384
Encompass_Preferred
The Human Glucagon (GC) ELISA Kit allows for quantitative determination of Glucagon in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 18.52-1,500 pg/mL. Sensitivity: 7.54 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P01275. Cross Activity: No significant cross-reactivity or interference between Glucagon and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2581