Learn More
Biomatik Corporation Chicken Immunoglobulin M (IgM) ELISA Kit
Supplier:  Biomatik Corporation EKU04910
Encompass_Preferred
Chicken Immunoglobulin M (IgM) ELISA Kit for detecting IgM in serum, plasma,, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 46.88-3,000pg/ml. Sensitivity: 19.27pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01875. Target Synonyms: IGHM; Immunoglobulin Heavy Constant Mu; Ig mu chain C region. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6093