Learn More
Biomatik Corporation Rat Glucagon (GC) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04386
Encompass_Preferred
Rat Glucagon (GCG) ELISA Kit for detecting GCG in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 18.52-1,500pg/ml. Sensitivity: 8.15pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P06883. Target Synonyms: GC; GLP1; GLP2; GRPP; Glicentin-Related Polypeptide; Glucagen; Oxyntomodulin; Incretin hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2583