Learn More
Biomatik Corporation Mouse Insulin Like Growth Factor 2 (IGF2) ELISA Kit
Supplier:  Biomatik Corporation EKU04977
Encompass_Preferred
Mouse Insulin Like Growth Factor 2 (IGF2) ELISA Kit for detecting IGF2 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: p09535. Gene ID: 16002. Target Synonyms: INSIGF; IGF-II; Somatomedin A; Preptin; Insulin-like growth factor II Ala-25 Del. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6097