Learn More
Biomatik Corporation Human Glucagon Like Peptide 1 Receptor (GLP1R) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04391
Encompass_Preferred
The Human Glucagon Like Peptide 1 Receptor (GLP1R) ELISA Kit allows for quantitative determination of Glucagon Like Peptide 1 Receptor in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.055 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P43220. Cross Activity: No significant cross-reactivity or interference between Glucagon Like Peptide 1 Receptor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2584