Learn More
Biomatik Corporation Human Insulin Like Growth Factor Binding Protein 1 (IGFBP1) ELISA Kit
Supplier:  Biomatik Corporation EKU04984
Encompass_Preferred
Human Insulin Like Growth Factor Binding Protein 1 (IGFBP1) ELISA Kit for detecting IGFBP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08833. Gene ID: 3484. Target Synonyms: IBP1; AFBP; IGF-BP25; PP12; Placental Protein 12; Amniotic Fluid Binding Protein; Alpha-Pregnancy-Associated Endometrial Globulin; Growth Hormone Independent-Binding Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6099