Learn More
Biomatik Corporation Mouse Glucagon Like Peptide 1 Receptor (GLP1R) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04392
Encompass_Preferred
Mouse Glucagon Like Peptide 1 Receptor (GLP1R) ELISA Kit for detecting GLP1R in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35659. Gene ID: 14652. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2585