Learn More
Biomatik Corporation Rat Insulin Like Growth Factor Binding Protein 2 (IGFBP2) ELISA Kit
Supplier:  Biomatik Corporation EKU04992
Encompass_Preferred
Rat Insulin Like Growth Factor Binding Protein 2 (IGFBP2) ELISA Kit for detecting IGFBP2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 62.5-4,000pg/ml. Sensitivity: 21.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P12843. Gene ID: 25662. Target Synonyms: IBP2; IGF-BP53. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6101