Learn More
Biomatik Corporation Human Glucagon Like Peptide 2 (GLP2) ELISA Kit, 96 tests
Catalog No. 501502586
Encompass_Preferred
The Human Glucagon Like Peptide 2 (GLP2) ELISA Kit allows for quantitative determination of Glucagon Like Peptide 2 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 123.5-10000 pg/mL. Sensitivity: 52.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Glucagon Like Peptide 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2586 Supplier Biomatik Corporation Supplier No. EKU04393
The Human Glucagon Like Peptide 2 (GLP2) ELISA Kit allows for quantitative determination of Glucagon Like Peptide 2 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 123.5-10000 pg/mL. Sensitivity: 52.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Glucagon Like Peptide 2 and analogues was observed. Restriction: For Research Use Only.