Learn More
Biomatik Corporation Human Integrin Alpha 6 (ITGa6) ELISA Kit
Supplier:  Biomatik Corporation EKU05035
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.054ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P23229. Target Synonyms: CD49f; CD49-F; ITG-A6; ITGA6B; VLA-6; CD49 antigen-like family member F; Processed integrin alpha-6. Restriction: For Research Use Only.
Catalog No. 50-228-6107