Learn More
Biomatik Corporation Mouse Glucokinase (GCK) ELISA Kit, 96 tests
Catalog No. 501502587
Encompass_Preferred
Mouse Glucokinase (GCK) ELISA Kit for detecting GCK in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P52792. Gene ID: 103988. Target Synonyms: GK; GLK; HHF3; HK4; HKIV; HXKP; MODY2; Hexokinase-D; Hexokinase 4; Maturity Onset Diabetes Of The Young 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2587 Supplier Biomatik Corporation Supplier No. EKU04401
Mouse Glucokinase (GCK) ELISA Kit for detecting GCK in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P52792. Gene ID: 103988. Target Synonyms: GK; GLK; HHF3; HK4; HKIV; HXKP; MODY2; Hexokinase-D; Hexokinase 4; Maturity Onset Diabetes Of The Young 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.