Learn More
Biomatik Corporation Simian Interferon Gamma (IFNg) ELISA Kit
Supplier:  Biomatik Corporation EKU05106
Encompass_Preferred
Simian Interferon Gamma (IFNg) ELISA Kit for detecting IFNg in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Simian. Detection Range: 7.8-500pg/ml. Sensitivity: 3.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P63310. Gene ID: 574282. Target Synonyms: IFN-G; IFG; IFI; INFr; IFN, Immune Interferon. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6116