Learn More
Biomatik Corporation Human Interleukin 1 Receptor Associated Kinase 1 (IRAK1) ELISA Kit
Supplier:  Biomatik Corporation EKU05161
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.113ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P51617. Target Synonyms: IRAK1; IRAK; Pelle. Restriction: For Research Use Only.
Catalog No. 50-228-6122