Learn More
Biomatik Corporation Rat Glucose Transporter 3 (GLUT3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04417
Encompass_Preferred
Rat Glucose Transporter 3 (GLUT3) ELISA Kit for detecting GLUT3 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q07647. Gene ID: 25551. Target Synonyms: SLC2A3; Solute Carrier Family 2 (Facilitated Glucose Transporter) Member 3; Glucose transporter type 3, brain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2593