Learn More
Biomatik Corporation Rat Interleukin 12A (IL12A) ELISA Kit
Supplier:  Biomatik Corporation EKU05204
Encompass_Preferred
Rat Interleukin 12A (IL12A) ELISA Kit for detecting IL12A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9R103. Gene ID: 84405. Target Synonyms: p35; IL12-A; CLMF1; NFSK; NKSF1; Natural Killer Cell Stimulatory Factor 1; Cytotoxic Lymphocyte Maturation Factor 1; NF Cell Stimulatory Factor Chain 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6130