Learn More
Biomatik Corporation Human Glucosidase Alpha, Acid (GaA) ELISA Kit, 96 tests
Catalog No. 501502598
Encompass_Preferred
The Human Glucosidase Alpha, Acid (GaA) ELISA Kit allows for quantitative determination of Glucosidase Alpha, Acid in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.067 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P10253. Cross Activity: No significant cross-reactivity or interference between Glucosidase Alpha, Acid and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2598 Supplier Biomatik Corporation Supplier No. EKU04425
The Human Glucosidase Alpha, Acid (GaA) ELISA Kit allows for quantitative determination of Glucosidase Alpha, Acid in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.067 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P10253. Cross Activity: No significant cross-reactivity or interference between Glucosidase Alpha, Acid and analogues was observed. Restriction: For Research Use Only.