Learn More
Biomatik Corporation Human Glucosidase Beta, Acid (GbA) ELISA Kit, 96 tests
Catalog No. 501502600
Encompass_Preferred
The Human Glucosidase Beta, Acid (GbA) ELISA Kit allows for quantitative determination of Glucosidase Beta, Acid in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2000 pg/mL. Sensitivity: 12.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P04062. Cross Activity: No significant cross-reactivity or interference between Glucosidase Beta, Acid and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2600 Supplier Biomatik Corporation Supplier No. EKU04427
The Human Glucosidase Beta, Acid (GbA) ELISA Kit allows for quantitative determination of Glucosidase Beta, Acid in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2000 pg/mL. Sensitivity: 12.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P04062. Cross Activity: No significant cross-reactivity or interference between Glucosidase Beta, Acid and analogues was observed. Restriction: For Research Use Only.