Learn More
Biomatik Corporation Rat Glucuronidase Beta (GUSb) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04431
Encompass_Preferred
Rat Glucuronidase Beta (GUSb) ELISA Kit for detecting GUSb in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.124ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06760. Gene ID: 24434. Target Synonyms: GUS-B; MPS7; Beta-G1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2601