Learn More
Biomatik Corporation Mouse Glutamate Cysteine Ligase, Catalytic (GCLC) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04432
Encompass_Preferred
Mouse Glutamate Cysteine Ligase, Catalytic (GCLC) ELISA Kit for detecting GCLC in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.119ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P97494. Gene ID: 14629. Target Synonyms: GCS; GLCL; GLCLC; Gamma-glutamylcysteine synthetase; Gamma-ECS. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2602