Learn More
Biomatik Corporation Human Interleukin 7 Receptor (IL7R) ELISA Kit
Supplier:  Biomatik Corporation EKU05372
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.057ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P16871. Target Synonyms: CDW127; CD127; IL-7R-Alpha. Restriction: For Research Use Only.
Catalog No. 50-228-6150