Learn More
Biomatik Corporation Rat Interleukin 8 Receptor Beta (IL8Rb) ELISA Kit
Supplier:  Biomatik Corporation EKU05389
Encompass_Preferred
Rat Interleukin 8 Receptor Beta (IL8Rb) ELISA Kit for detecting IL8Rb in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 10.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35407. Gene ID: 29385. Target Synonyms: CD182; CDw128b; IL8R-B; IL8-RB; CMKAR2; CXCR2; IL8R2; C-X-C chemokine receptor type 2; GRO/MGSA receptor; High affinity interleukin-8 receptor B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6153