Learn More
Biomatik Corporation Human Glutamate Decarboxylase 2, Acid (GAD2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04439
Encompass_Preferred
The Human Glutamate Decarboxylase 2, Acid (GAD2) ELISA Kit allows for quantitative determination of Glutamate Decarboxylase 2, Acid in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.27 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q05329. Cross Activity: No significant cross-reactivity or interference between Glutamate Decarboxylase 2, Acid and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2604