Learn More
Biomatik Corporation Mouse Glutamate Dehydrogenase 1 (GDH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04443
Encompass_Preferred
Mouse Glutamate Dehydrogenase 1 (GLUD1) ELISA Kit for detecting GLUD1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.072ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P26443. Gene ID: 14661. Target Synonyms: GDH; Glutamate dehydrogenase 1, mitochondrial. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2605